Sulfuric acid 9.5 - 10.5% (w/v) R.S., Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

MACRH378-05 MACRH378-07 MACRH378-09
CAMKH37805EA 104.96 CAD
CAMKH37805 CAMKH37807 CAMKH37809
Sulfuric acid 9.5 - 10.5% (w/v) R.S., Macron Fine Chemicals™
Sulfuric acid
Formula: H₂SO₄
Boiling Pt: 101 °C (1013 hPa)
Density: 1.066 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II
Merck Index: 13,09064

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (H₂SO₄)(g/100 mL) 9.5 - 10.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR