Volumetric Flasks with [ST] Glass Stopper, Class A, Serialized and Certified, Hirschmann
Supplier: Hirschmann
282-G-10S 282-G-25S 282-G-50S 282-G-100S 282-G-250S 282-G-500S 282-G-1000S 282-G-2000S
89025-764PK 277.56 CAD
89025-764 89025-766 89025-768 89025-770 89025-774 89025-776 89025-778 89025-780
Volumetric Flasks with [ST] Glass Stopper, Class A, Serialized and Certified, Hirschmann
Flasks Volumetric Flasks
Flasks feature a fine white graduation line, serial number, and marking spot
Meet ASTM E288, Class A requirements.
Graduated to contain. Flasks include a [ST] glass stopper and a certification of identification and accuracy.
Order Now
SPECIFICATIONS
Environmentally Preferable