Potassium dihydrogen phosphate, ULTREX®, Ultrapure

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

Management of Change (MOC) category = N

BAKR4921-04 BAKR4921-07
CAJT4921-4EA 938.68 CAD
CAJT4921-4 CAJT4921-7
Potassium dihydrogen phosphate, ULTREX®, Ultrapure
Potassium dihydrogen phosphate

Certificate of lot analysis provided.

Formula: KH₂PO₄
MW: 136.09 g/mol
Boiling Pt: ~100 °C (1013 hPa)
Melting Pt: 253
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (KH₂PO₄) (dried basis), For Information Only
Loss on Drying at 105°C, For Information Only
Particulate Matter, For Information Only
pH of 5% Solution at 25°C, For Information Only
Non-Metallic Impurities - Arsenic (As), For Information Only
Non-Metallic Impurities - Fluoride (F), For Information Only
Non-Metallic Impurities - Halide (as Cl), For Information Only
Non-Metallic Impurities - Nitrogen Compounds (as N), For Information Only
Non-Metallic Impurities - Silicon (Si), For Information Only
Non-Metallic Impurities - Sulfate (SO₄), For Information Only
Metallic Impurities - Aluminum (Al), For Information Only
Metallic Impurities - Barium (Ba), For Information Only
Metallic Impurities - Bismuth (Bi), For Information Only
Metallic Impurities - Cadmium (Cd), For Information Only
Metallic Impurities - Calcium (Ca), For Information Only
Metallic Impurities - Chromium (Cr), For Information Only
Metallic Impurities - Cobalt (Co), For Information Only
Metallic Impurities - Copper (Cu), For Information Only
Metallic Impurities - Iron (Fe), For Information Only
Metallic Impurities - Lead (Pb), For Information Only
Metallic Impurities - Magnesium (Mg), For Information Only
Metallic Impurities - Manganese (Mn), For Information Only
Metallic Impurities - Mercury (Hg), For Information Only
Metallic Impurities - Molybdenum (Mo), For Information Only
Metallic Impurities - Nickel (Ni), For Information Only
Metallic Impurities - Niobium (Nb), For Information Only
Metallic Impurities - Silver (Ag), For Information Only
Metallic Impurities - Sodium (Na), For Information Only
Metallic Impurities - Strontium (Sr), For Information Only
Metallic Impurities - Tin (Sn), For Information Only
Metallic Impurities - Titanium (Ti), For Information Only
Metallic Impurities - Vanadium (V), For Information Only
Metallic Impurities - Zinc (Zn), For Information Only
Metallic Impurities - Zirconium (Zr), For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 400 nm, For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 280 nm, For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 254 nm, For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 220 nm, For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 215 nm, For Information Only
Certificate Provided Reports Actual Lot Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR