Whizard® Polyester/Wire Cut-Resistant Glove Liners, Wells Lamont
Supplier: Wells Lamont
M281S M281M M281L M281XL
32935-392BG 392.88 CAD
32935-392 32935-394 32935-396 32935-398
Whizard® Polyester/Wire Cut-Resistant Glove Liners, Wells Lamont
Gloves
Wire cut-resistant glove liners are made from polyester fiber and stainless steel wire to provide the ultimate cut resistance.
  • Ambidextrous
  • Cuff color denotes sizes
A true lightweight alternative to steel mesh gloves, the reusable black gloves feature ergonomic design to offer comfort without sacrificing cut resistance.
Certifications: Can be used as outside glove in ISO Class 5 (FED-STD-209E Class 100/M3.5) conditions.