Volumetric Flasks with [ST] Polyethylene Stopper, Class A, Hirschmann
Supplier: Hirschmann
282-P-5 282-P-10 282-P-50 282-P-100 282-P-200 282-P-250 282-P-500 282-P-1000 282-P-2000
89025-798PK 151.66 CAD
89025-798 89025-800 89025-804 89025-806 89025-808 89025-810 89025-812 89025-814 89025-816
Volumetric Flasks with [ST] Polyethylene Stopper, Class A, Hirschmann
Flasks Volumetric Flasks
Flasks feature a fine white graduation line and marking spot
Meet ASTM E288, Class A requirements.
Graduated to contain. Flasks include a [ST] polyethylene stopper.