VWR® Type K Thermocouple Probes
Supplier: VWR International
23609-238 61161-305 77776-728 89030-232 61161-372
23609-238EA 468.55 CAD
23609-238 61161-305 77776-728 89030-232 61161-372
VWR® Type K Thermocouple Probes
Temperature Probes
For use with all Type-K thermometers in liquids, semi-solids, and air/gas.
Probe 61161-372 has fluoropolymer resin insulation. For improved performance, probes should be calibrated as a system with the meter.
Note: Type K probes are not ISO/IEC 17025 accredited