Volumetric Flasks with [ST] Glass Stopper, Class A, Hirschmann
Supplier: Hirschmann
282-G-5 282-G-10 282-G-25 282-G-50 282-G-100 282-G-200 282-G-250 282-G-500 282-G-1000 282-G-2000
89025-744PK 167.91 CAD
89025-744 89025-746 89025-748 89025-750 89025-752 89025-754 89025-756 89025-758 89025-760 89025-762
Volumetric Flasks with [ST] Glass Stopper, Class A, Hirschmann
Flasks Volumetric Flasks
Flasks feature a fine white graduation line and marking spot
Meet ASTM E288, Class A requirements.
Graduated to contain. Flasks include a [ST] glass stopper.