Sodium sulfate, anhydrous ≥99.0%, granular, AR® ACS 10 - 60 Mesh, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt

8024-06 MACR8024-24 MACR8024-20 MACR8024-04
MK802406CS 1453.21 CAD
MK802406 CAMK802404 CAMK802406 CAMK802424 CAMK802420 CAJT8024-QM CAMK8024-04
Sodium sulfate, anhydrous ≥99.0%, granular, AR® ACS 10 - 60 Mesh, Macron Fine Chemicals™
Sodium sulphate

AR—The standard Macron Fine Chemicals™ grade of analytical reagents; suitable for laboratory and general use. If the reagent also meets the requirements of the American Chemical Society Committee on Analytical Reagents, it will be denoted as an AR (ACS) reagent. 


AR—The standard Macron Fine Chemicals™ grade of analytical reagents; suitable for laboratory and general use. If the reagent also meets the requirements of the American Chemical Society Committee on Analytical Reagents, it will be denoted as an AR (ACS) reagent. 

Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429
Melting Pt: 888
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Appearance (white, crystalline granules) Passes Test
ACS - Assay (Na₂SO₄) ≥99.0 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.01 %
ACS - Iron (Fe) ≤0.001 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Loss on Ignition ≤0.5 %
ACS - Nitrogen Compounds (as N) ≤5 ppm
ACS - pH of 5% Solution at 25°C 5.2 - 9.2
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Potassium (K) ≤0.01 %
Solubility and Foreign Matter Passes Test
On U.S. No. 10 Sieve ≤1 %
On U.S. No. 60 Sieve ≥80 %
Thru U.S. No. 60 Sieve ≤19 %
Thru U.S. No. 100 Sieve ≤10 %
Extraction - Concentration Suitability Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR