PIPES ≥99.0%, Ultrapure Bioreagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

BAKR4265-04 BAKR4265-01
CAJT4265-4EA 2181.9 CAD
CAJT4265-4 CAJT4265-1
PIPES ≥99.0%, Ultrapure Bioreagent, J.T.Baker®
PIPES

For molecular biology applications. DNase-, RNase-, and protease-free.

Formula: C₈H₁₈N₂O₆S₂
Boiling Pt: 499 °C (1013 hPa)
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006159
CAS Number: 5625-37-6
EINECS: 227-057-6
Merck Index: 12,07633
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5192468b680c7bea85a31a131811618356a531b9&vid=ef0e3d488c68ea7f6f583fede6ebf7d2196392b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₈H₁₈N₂O₆S₂) ≥99.0 %
Appearance Passes Test
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test
ACS - Heavy Metals (as Pb) ≤5 ppm
Insoluble Matter ≤0.005 %
Residue after Ignition ≤0.30 %
Product Information (not specifications): pKa at 20°C = 6.80


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR