Potassium dihydrogen phosphate ≥99.0%, crystals, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
prim.-Potassium phosphate
, Monopotassiumphosphate
, Potassium dihydrogen orthophosphate
, Potassium phosphate monobasic
, Monobasic Potassium Phosphate
, Monopotassium phosphate
Management of Change (MOC) category = NI
BAKR4008-05
BAKR4008-01
4008-01
4008-05
CAJT4008-1EA
2530.31
CAD
CAJT4008-1
CAJT4008-5
JT4008-1
JT4008-5
Potassium dihydrogen phosphate ≥99.0%, crystals, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Potassium dihydrogen phosphate
Crystals. For LC and molecular biology. Lot analysis on label.
Formula:
KH₂PO₄ MW: 136.09 g/mol Boiling Pt: ∼100 °C (1013 hPa) Melting Pt: 253 °C Density: 2.3 g/cm³ (20 °C) |
MDL Number:
MFCD00011401 CAS Number: 7778-77-0 Merck Index: 13,07744 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (KH₂PO₄) | ≥99.0 % |
DNase Activity (None Detected) | Passes Test |
RNase Activity | Passes Test |
Protease Activity | Passes Test |
pH of 5% Solution at 25°C | 4.1 - 4.5 |
Insoluble Matter, Calcium and NH₄OH Precipitate | ≤0.01 % |
Loss on Drying at 105°C | ≤0.2 % |
Heavy Metals (as Pb) | ≤0.001 % |
Iron (Fe) | ≤0.002 % |
Trace Impurities - Arsenic (As) | ≤3.000 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...