Sodium pyruvate ≥99%, white powder, BAKER ANALYZED®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

Management of Change (MOC) category = N

BAKR3354-02 BAKR3354-04
CAJT3354-2EA 2011.8 CAD
CAJT3354-2 CAJT3354-4
Sodium pyruvate ≥99%, white powder, BAKER ANALYZED®
Sodium pyruvate

Lot analysis on label.

Formula: CH₃COCOONa
Storage Temperature: Refrigerator
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃COCOONa) (by non-aqueous titration) ≥99.0 %
Free Pyruvic Acid (CH₃COCOOH) ≤1 %
Solubility Passes Test
Meets Reagent Specifications for testing USP/NF monographs.
Store cold. Keep Dry. Warm to room temperature before opening.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR