Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder, GenAR® USP for biotechnology, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = R

MACR7749-88 MACR7749-06 MACR7749-24
CAMK774988EA 575.91 CAD
CAMK774988 CAMK7749-06 CAMK7749-24
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder, GenAR® USP for biotechnology, Macron Fine Chemicals™
Sodium hydrogen carbonate

Avantor excipients are used during the formulation process in a wide range of applications. Avantor provides high-purity and performance excipients that serve as fillers, binders, disintegrants, lubicants, flavors, emulsifiers and preservatives. All the products are cGMP produced and meet USP grade standards.

Formula: NaHCO₃
Melting Pt: 270 °C
Density: 2.159 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Food GMP Manufactured Product
GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
USP - Limit of Ammonia ≤ 20 ppm
USP - Limit of Sulfur Compounds ≤ 0.015 %
USP - Arsenic (As) ≤ 2 ppm
USP - Normal Carbonate Passes Test
USP - Chloride (Cl) ≤ 0.015 %
USP - Identification A (Sodium) Passes Test
USP - Identification B (Bicarbonate) Passes Test
USP - Insoluble Substances Passes Test
USP - Loss on Drying ≤ 0.25 %
EP/BP - Assay (NaHCO₃) 99.0 - 101.0 %
EP/BP - Ammonium (NH₄) ≤ 20 ppm
EP/BP - Appearance of Solution Passes Test
EP/BP - Calcium (Ca) ≤ 100 ppm
EP/BP - Carbonate (pH) ≤ 8.6
EP/BP - Chloride (Cl) ≤ 150 ppm
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Iron (Fe) ≤ 20 ppm
EP/BP - Sulfate (SO₄) ≤ 150 ppm
JP - Assay (NaHCO₃) 99.0 - 101.0 %
JP - Identification Passes Test
JP - pH (1 in 20) 7.9 - 8.4
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤ 0.040 %
JP - Carbonate (CO₃) Passes Test
JP - Ammonium (NH₄) Passes Test
JP - Heavy Metals (as Pb) ≤ 5 ppm
JP - Arsenic (As) ≤ 2 ppm
FCC - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
FCC - Insoluble Substances Passes Test
FCC - Lead (Pb) ≤ 2 mg/kg
FCC - Limit of Ammonia (NH₃) Passes Test
Endotoxin Concentration (EU/g) ≤ 5.0
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR