Highly Reusable Nylon Glove Liners, Wells Lamont
Supplier: Wells Lamont
M555S M555M M555L M555XL
32935-581BG 128.52 CAD
32935-581 32935-583 32935-585 32935-587
Highly Reusable Nylon Glove Liners, Wells Lamont
Gloves
Thin, conformable nylon glove liners provide excellent fit, feel, and comfort.
  • Ambidextrous
  • Full-finger style
  • Regular cuff
  • Gloves prevent skin irritation
  • Nylon
The liners are designed as a protective barrier between user's hands and latex gloves to prevent skin irritation without affecting tactile sensitivity.
Ambidextrous, full-finger style gloves feature a regular cuff. Cuff color denotes size.
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable