Sodium carbonate, anhydrous ≥99.95%, granules, AR® ACS primary standard, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
Calcined soda, Carbonic acid disodium salt
Management of Change (MOC) category = N
MACR7528-20
MACR7528-04
CAMK752820EA
3038.1
CAD
CAMK752820
CAMK7528-04
Sodium carbonate, anhydrous ≥99.95%, granules, AR® ACS primary standard, Macron Fine Chemicals™
Sodium carbonate
Granular. Primary standard.
Formula:
Na₂CO₃ MW: 105.99 g/mol Boiling Pt: 1600 Melting Pt: 854 Density: 2.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003494 CAS Number: 497-19-8 EINECS: 207-838-8 REACH: 01-2119485498-19 Merck Index: 14,08596 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (Na₂CO₃ (after 2 hr @ 285°C)) | 99.95 - 100.05 % |
ACS - Calcium (Ca) | ≤0.02 % |
ACS - Chloride (Cl) | ≤0.001 % |
ACS - Heavy Metals (as Pb) | ≤0.0005 % |
ACS - Insoluble Matter | ≤0.01 % |
ACS - Iron (Fe) | ≤0.0005 % |
ACS - Loss on Heating at 285°C | ≤1.0 % |
ACS - Magnesium (Mg) | ≤0.004 % |
ACS - Phosphate (PO₄) | ≤0.001 % |
ACS - Potassium (K) | ≤0.005 % |
ACS - Silica (SiO₂) | ≤0.005 % |
ACS - Sulfur Compounds (as SO₄) | ≤0.003 % |
ACS - Solubility (8 in 50) | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...