Sodium carbonate monohydrate ≥99.5%, granular, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
MACR7516-04
MACR7516-06
CAMK751604CS
2488.11
CAD
CAMK751604
CAMK751606
Sodium carbonate monohydrate ≥99.5%, granular, AR® ACS, Macron Fine Chemicals™
Sodium carbonate monohydrate
Granular.
Formula:
Na₂CO₃·1H₂O Melting Pt: 107.85 °C Density: 2.25 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149177 CAS Number: 5968-11-6 EINECS: 207-838-8 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (Na₂CO₃·H₂O) (by acidimetry) | ≥99.5 % |
ACS - Loss on Drying at 150°C | 13.0 - 15.0 % |
ACS - Insoluble Matter | ≤0.01 % |
ACS - Chloride (Cl) | ≤0.001 % |
ACS - Phosphate (PO₄) | ≤5 ppm |
ACS - Silica (SiO₂) | ≤0.005 % |
ACS - Sulfur Compounds (as SO₄) | ≤0.004 % |
ACS - Heavy Metals (as Pb) | ≤5 ppm |
ACS - Iron (Fe) | ≤5 ppm |
ACS - Calcium (Ca) | ≤0.03 % |
ACS - Magnesium (Mg) | ≤0.005 % |
ACS - Potassium (K) | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...