Sodium bisulfite ≥58.5%, granular, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Sodium hydrogen sulphite, Sodium hydrogen sulfite

Management of Change (MOC) category = N

7448-06 MACR7448-06 MACR7448-12 MACR7448-20 MACR7448-24
MK744806CS 2542.28 CAD
MK744806 CAMK744806 CAMK744812 CAMK744820 CAMK744824
Sodium bisulfite ≥58.5%, granular, AR® ACS, Macron Fine Chemicals™
Sodium hydrogen sulphite

Granular. A mixture of sodium bisulfite (NaHSO3) and sodium metabisulfite (Na2S2O5).

Formula: HNaO₃S
MW: 104.06 g/mol
Melting Pt: 300 °C
Density: 2.476 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7631-90-5
EINECS: 231-548-0
UN: 2693
ADR: 8,III
Merck Index: 12,08731

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (as SO₂) ≥58.5 %
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤0.02 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR