Sodium bisulfite ≥90% FCC, purified, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Sodium hydrogen sulphite, Sodium hydrogen sulfite

Management of Change (MOC) category = RL

7444-03 7444-06 MACR7444-03 MACR7444-06 MACR7444-20
MK744403EA 108.75 CAD
MK744403 MK744406 CAMK744403 CAJT7444-QM CAMK744406 CAMK744420
Sodium bisulfite ≥90% FCC, purified, Macron Fine Chemicals™
Sodium hydrogen sulphite

Granular. A mixture of sodium bisulfite (NaHSO₃) and sodium metabisulfite (Na₂S₂O₅).

Formula: HNaO₃S
MW: 104.06 g/mol
Melting Pt: 300
Density: 2.476 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7631-90-5
EINECS: 231-548-0
UN: 2693
ADR: 8 III
Merck Index: 12,08731

Order Now

Specification Test Results

Meets F.C.C. Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
FCC - Assay (as (Na₂S₂O₅)) ≥90 %
FCC - Assay (SO₂) 58.5 - 67.4 %
Solubility Passes Test
FCC - Identification A Passes Test
FCC - Identification B Passes Test
Insoluble Matter ≤0.005 %
Thiosulfate (S₂O₃) ≤0.05 %
Arsenic (As) ≤1 ppm
FCC - Lead (Pb) ≤2 mg/kg
FCC - Iron (Fe) ≤0.005 %
FCC - Selenium (Se) ≤5 mg/kg
Chloride (Cl) ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR