Sodium hydrogen carbonate 99.0-100.5% (by acidimetry on anhydrous substance), powder USP, FCC for haemodialysis, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium bicarbonate

Management of Change (MOC) category = R

MACR7396-04 7396-88 MACR7396-08 MACR7396-06 MACR7396-88 MACR7396-26 MACR7396-20 7396-06 MACR7396-08 7396-26 7396-20 7396-08
CAMK739606EA 759.77 CAD
CAMK739606 CAJT7396-08 CAMK739626 MK739620 MK739688 JT7396-08 MK7396-08 CAMK739620 MK739604 MK739626 MK739606 CAMK739688
Sodium hydrogen carbonate 99.0-100.5% (by acidimetry on anhydrous substance), powder USP, FCC for haemodialysis, Macron Fine Chemicals™
Sodium hydrogen carbonate

pH adjusters are used to adjust the pH of formulations.

Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270 °C
Density: 2.159 g/cm³ (20 °C)
MDL Number: MFCD00003528
CAS Number: 144-55-8
Merck Index: 13,08655
https://ca.vwr.com/stibo/web/std.lang.all/05/81/16640581.jpg

Order Now

Specification Test Results

Meets F.C.C. Requirements
GMP Manufactured Product
Meets U.S.P Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (NaHCO₃) (dried basis)(by acidimetry) 99.0 - 100.5 %
USP - Aluminum (Al) ≤2 ppm
USP - Arsenic (As) ≤2 ppm
USP - Calcium (Ca) ≤0.01 %
USP - Carbonate (CO₃) ≤0.23 %
USP - Chloride (Cl) ≤0.015 %
USP - Copper (Cu) ≤1 ppm
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Substances Passes Test
USP - Iron (Fe) ≤5 ppm
USP - Limit of Ammonia ≤20 ppm
USP - Limit of Sulfur Compounds ≤0.015 %
USP - Loss on Drying ≤0.25 %
USP - Magnesium (Mg) ≤0.004 %
USP - Normal Carbonate Passes Test
USP - Limit of Organics ≤0.01 %
FCC - Assay (NaHCO₃) (dried basis)(by acidimetry) 99.0 - 100.5 %
FCC - Ammonia (as NH₃) Passes Test
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Insoluble Substances Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Drying ≤0.25 %
The net weight of the product in each container is within +/- 2% of the
netweight stated on the label.
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR