Sodium acetate, anhydrous ≥99.0%, white powder, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = NI

MACR7372-05 MACR7372-12 MACR7372-20 MACR7372-24
CAMK737205EA 1266.78 CAD
CAMK737205 CAMK737212 CAMK737220 CAMK737224
Sodium acetate, anhydrous ≥99.0%, white powder, AR® ACS, Macron Fine Chemicals™
Sodium acetate

Powder.

Formula: H₃CCOONa
Boiling Pt: 881.4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1.528 g/cm³ (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃COONa) ≥99.0 %
ACS - pH of 5% Solution at 25°C 7.0 - 9.2
ACS - Insoluble Matter ≤0.01 %
ACS - Loss on Drying at 120°C ≤1.0 %
ACS - Chloride (Cl) ≤0.002 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Sulfate (SO₄) ≤0.003 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Magnesium (Mg) ≤0.002 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤0.001 %
Appearance (fine, white powder)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR