Sand, dried, washed and dried, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

MACR7062-06
MK706206CS 1586.54 CAD
MK706206
Sand, dried, washed and dried, Macron Fine Chemicals™
Sand

Sand washed and dried.


Caution: For laboratory, research, or manufacturing use.

Formula: SiO₂
MW: 60.08 g/mol
Melting Pt: 1427 °C
MDL Number: MFCD02100519
CAS Number: 7631-86-9

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Substances Soluble in HCl ≤0.16 %
Appearance (fine, off-white to gray granules)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR