Volumetric Flasks, Mixing Bulb, Class A, Chemglass
Supplier: Chemglass
CG-1601-08 CG-1601-04 CG-1601-06 CG-1601-07 CG-1601-09
80070-380EA 361.65 CAD
80070-380 80070-372 80070-376 80070-378 80070-382
Volumetric Flasks, Mixing Bulb, Class A, Chemglass
Flasks Volumetric Flasks
Flasks feature a mixing bulb between the calibration line and stopper to aid in the mixing or dissolving of the contents and a solid glass pennyhead stopper
Conforms to Federal Specifications NNN-F-289 for Type 1, Class A unserialized flasks and ASTM E288.
The mixing bulb contains approximately one-tenth of the flask volume.