Silica gel, SilicAR Grade 62, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Management of Change (MOC) category = N
MACR6551-02
MACR6551-05
CAMK655102CS
2107.75
CAD
CAMK655102
CAMK655105
Silica gel, SilicAR Grade 62, Macron Fine Chemicals™
Silica gel
60 to 200 mesh. For column chromatography.
Formula:
SiO₂ |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Loss on Ignition | ≤12 % |
On U.S. No. 60 Sieve | ≤5 % |
On U.S. No. 200 Sieve | ≥90 % |
Appearance (fine, white powder) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...