Sodium hydroxide 0.10 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Caustic soda

Management of Change (MOC) category = NI

MACR6146-01 MACR6146-09 MACR6146-60
CAMK614601EA 210.66 CAD
CAMK614601 CAMK614609 CAMK614660
Sodium hydroxide 0.10 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 III
Merck Index: 14,08627

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Protect from air to avoid absorption of carbon dioxide.
Normality 0.0995 - 0.1005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR