Silver nitrate 0.10 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

Management of Change (MOC) category = NI

MACR6142-01 MACR6142-60
CAMK614201EA 1020.29 CAD
CAMK614201 CAMK614260
Silver nitrate 0.10 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.095 - 0.105

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR