Sodium thiosulfate 0.1 N (N/10) aqueous solution, StandARd® Volumetric, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

M.O.C. Category NI

4682-60 MACR4682-01
MK468260EA 76.33 CAD
MK468260 CAMK468201
Sodium thiosulfate 0.1 N (N/10) aqueous solution, StandARd® Volumetric, Macron Fine Chemicals™
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR