Sulfuric acid 95.0-98.0%, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

MACR2876-05 MACR2876-18 MACR2876-45 MACR2876-46 MACR2876-28 MACR2876-14 MACR2876-21
CAMK287605CS 1014.71 CAD
CAMK287605 CAMK287618 CAMK287645 CAMK287646 CAJT2876-14 CAJT2876-21 CAMK287628 MK2876-14 MK2876-21
Sulfuric acid 95.0-98.0%, AR® ACS, Macron Fine Chemicals™
Sulfuric acid

ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  

Formula: H₂SO₄
MW: 98.08
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
ACS - Assay (H₂SO₄) 95.0 - 98.0 %
ACS - Color (APHA) ≤10
ACS - Residue after Ignition ≤5 ppm
ACS - Chloride (Cl) ≤0.2 ppm
ACS - Nitrate (NO₃) ≤0.5 ppm
ACS - Ammonium (NH₄) ≤2 ppm
ACS - Substances Reducing Permanganate (as SO₂) ≤2 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Mercury (Hg) ≤5.0 ppb
Trace Impurities - Arsenic (As) ≤0.010 ppm
Trace Impurities - Boron (B) ≤0.010 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR