di-Sodium L(+)-tartrate dihydrate 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium L-(+)-tartrate dihydrate, L(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Sodium tartrate dibasic dihydrate, Disodium L-(+)-Tartrate Dihydrate, (R,R)-(+)-Disodium tartrate dihydrate, L(+)-Tartaric acid, disodium salt, dihydrate, Sodium L-tartrate dibasic dihydrate, L(+)-Sodium tartrate dibasic dihydrate, Sodium Tartrate, Dihydrate, (+)-Disodium L-tartrate dihydrate, (+)-Disodium-L-tartrat dihydrate

Management of Change (MOC) category = N

2386-04 MACR2386-20
MK238604CS 4932.43 CAD
MK238604 CAMK238620
di-Sodium L(+)-tartrate dihydrate 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™
di-Sodium L(+)-tartrate dihydrate

Granular.

Formula: NaOOC(CHOH)₂COONa·2H₂O
MW: 230.08 g/mol
Melting Pt: 57
Density: 1.818 g/cm³ (20 °C)
MDL Number: MFCD00150035
CAS Number: 6106-24-7
EINECS: 212-773-3
Merck Index: 12,08833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39a75038f4ff5e6c234267b15540823ae361bed4&vid=89289572c48b0c6415f8f11e916a51f66e129474&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Na₂C₄H₄O₆·2H₂O) 99.0 - 101.0 %
ACS - Loss on Drying at 150°C 15.61 - 15.71 %
ACS - pH of 5% Solution at 25°C 7.0 - 9.0
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤5 ppm
ACS - Phosphate (PO₄) ≤5 ppm
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Ammonium (NH₄) ≤0.003 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Iron (Fe) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR