Silver nitrate ≥99.0%, crystals, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

MACR2169-03 MACR2169-34
CAMK216901CS 14880.12 CAD
CAMK216901 CAMK216903 CAMK216934
Silver nitrate ≥99.0%, crystals, AR® ACS, Macron Fine Chemicals™
Silver nitrate

Crystals.

Formula: AgNO₃
Boiling Pt: 444 °C (1013 hPa)
Melting Pt: 210 °C
Density: 4.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (AgNO₃) ≥99.0 %
ACS - Clarity of Solution Passes Test
ACS - Chloride (Cl) ≤5 ppm
ACS - Free Acid Passes Test
ACS - Substances not Precipitated by HCl ≤0.01 %
ACS - Sulfate (SO₄) ≤0.002 %
ACS - Copper (Cu) ≤2 ppm
ACS - Iron (Fe) ≤2 ppm
ACS - Lead (Pb) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR