Sodium azide ≥99.7%, OR, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

1953-59 MACR1953-09
MK195359EA 367.63 CAD
MK195359 CAMK195309
Sodium azide ≥99.7%, OR, Macron Fine Chemicals™
Sodium azide

Macron Fine Chemicals™ OR: organic reagents that are suitable for research applications.

Formula: NaN₃
Melting Pt: 275 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003536
CAS Number: 26628-22-8
EINECS: 247-852-1
UN: 1687
ADR: 6.1,II
Merck Index: 13,08653

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (NaN₃) ≥99.7 %
Loss on Drying at 105°C ≤0.1 %
Typical property (not a specification): Appearance (white, crystalline
powder) White, Crystalline Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR