tri-Sodium citrate dihydrate 99 Anhydrous basis, crystals FCC, USP, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

M.O.C. Category R

MACR0734-06 MACR0734-23 MACR0734-88 MACR0734-04
CAMK073406CS 1482.34 CAD
CAMK073406 CAMK073404 CAMK073423 CAMK073488 CAMK0734-04
tri-Sodium citrate dihydrate 99 Anhydrous basis, crystals FCC, USP, Macron Fine Chemicals™
tri-Sodium citrate dihydrate

Colorless or white to slightly yellow crystals. GMP manufactured.

Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (anhydrous basis) 99.0 - 100.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - Water (H₂O) 10.0 - 13.0 %
USP - Alkalinity Passes Test
USP - Tartrate Passes Test
FCC - Assay (anhydrous basis) 99.0 - 100.5 %
FCC - Identification Passes Test
FCC - Alkalinity Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Water (H₂O) 10.0 - 13.0 %
Thru U.S. No. 50 Sieve, For Information Only
Solubility (5 in 50) Passes Test
Appearance (colorless or white to slightly yellow crystals)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR