Sodium dodecyl sulfate (SDS) ≥95%, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium dodecyl sulfateLauryl sulfate sodium saltSodium lauryl sulfateDodecyl sulfate sodium saltDodecyl sulphuric acid sodium saltLauryl sulphate sodium saltSodium lauryl sulphateDodecyl sulphate sodium saltDodecyl sulfuric acid sodium salt

Management of Change (MOC) category = N

BAKRL050-02 L050-07 BAKRL050-07
JTL050-02CS 419.74 CAD
JTL050-02 CAJTL050-7 JTL050-7
Sodium dodecyl sulfate (SDS) ≥95%, J.T.Baker®
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
MDL Number: MFCD00036175
CAS Number: 151-21-3
UN: 2926
ADR: 4.1, 6.1,II
Merck Index: 14,08636
https://ca.vwr.com/stibo/web/std.lang.all/54/91/735491.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (as C₁₂H₂₅OSO₃Na) ≥95 %
Identification (by IR) Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR