Sulfuric acid ≥95%, BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

BAKR9681-01 BAKR9681-15 BAKR9681-08 BAKR9681-00 9681-05 9681-07 9681-02 9681-03
CAJT9681-1CS 1180.53 CAD
CAJT9681-1 CAJT9681-15 CAJT9681-8 CAJT9681-0 JT9681-5 JT9681-7 JT9681-2 JT9681-3 CAJT9681-69
Sulfuric acid ≥95%, BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®
Sulfuric acid

The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.

Formula: H₂SO₄
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (H₂SO₄) 95.0 - 98.0 %
Appearance Passes Test
Color (APHA) ≤10
Specific Gravity at 60°/60°F ≥1.84
Residue after Ignition ≤4 ppm
Ammonium (NH₄) ≤1 ppm
Trace Impurities - Arsenic (As) ≤0.004 ppm
Chloride (Cl) ≤0.1 ppm
Nitrate (NO₃) ≤0.5 ppm
Trace Impurities - Aluminum (Al) ≤200.0 ppb
Trace Impurities - Calcium (Ca) ≤200.0 ppb
Trace Impurities - Chromium (Cr) ≤100.0 ppb
Trace Impurities - Copper (Cu) ≤100.0 ppb
Trace Impurities - Gold (Au) ≤100.0 ppb
Heavy Metals (as Pb) ≤500.0 ppb
Trace Impurities - Iron (Fe) ≤200.0 ppb
Trace Impurities - Lead (Pb) ≤200.0 ppb
Trace Impurities - Magnesium (Mg) ≤200.0 ppb
Trace Impurities - Manganese (Mn) ≤100.0 ppb
Trace Impurities - Mercury (Hg) ≤5.0 ppb
Trace Impurities - Nickel (Ni) ≤200.0 ppb
Trace Impurities - Potassium (K) ≤300.0 ppb
Trace Impurities - Sodium (Na) ≤300 ppb
Trace Impurities - Tin (Sn) ≤200.0 ppb
Trace Impurities - Titanium (Ti) ≤200.0 ppb
Trace Impurities - Zinc (Zn) ≤200.0 ppb
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR