Sulfuric acid ≥95%, BAKER INSTRA-ANALYZED® for trace metal analysis low Selenium, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

BAKR9673-00 BAKR9673-33
CAJT9673-0CS 1093.17 CAD
CAJT9673-0 CAJT9673-33
Sulfuric acid ≥95%, BAKER INSTRA-ANALYZED® for trace metal analysis low Selenium, J.T.Baker®
Sulfuric acid

The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.

Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (H₂SO₄) 95.0 - 98.0 %
Appearance Passes Test
ACS - Color (APHA) ≤10
ACS - Residue after Ignition ≤3 ppm
ACS - Substances Reducing Permanganate (as SO₂) ≤2 ppm
Ammonium (NH₄) ≤1 ppm
Chloride (Cl) ≤0.1 ppm
Nitrate (NO₃) ≤0.2 ppm
Phosphate (PO₄) ≤0.5 ppm
Trace Impurities - Aluminum (Al) ≤30.0 ppb
Arsenic and Antimony (as As) ≤4.0 ppb
Trace Impurities - Barium (Ba) ≤10.0 ppb
Trace Impurities - Beryllium (Be) ≤10.0 ppb
Trace Impurities - Bismuth (Bi) ≤10.0 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤2.0 ppb
Trace Impurities - Calcium (Ca) ≤50.0 ppb
Trace Impurities - Chromium (Cr) ≤6.0 ppb
Trace Impurities - Cobalt (Co) ≤0.5 ppb
Trace Impurities - Copper (Cu) ≤1.0 ppb
Trace Impurities - Gallium (Ga) ≤10.0 ppb
Trace Impurities - Germanium (Ge) ≤10.0 ppb
Trace Impurities - Gold (Au) ≤10.0 ppb
Heavy Metals (as Pb) ≤500.0 ppb
Trace Impurities - Iron (Fe) ≤50.0 ppb
Trace Impurities - Lead (Pb) ≤0.5 ppb
Trace Impurities - Lithium (Li) ≤10.0 ppb
Trace Impurities - Magnesium (Mg) ≤7.0 ppb
Trace Impurities - Manganese (Mn) ≤1.0 ppb
Trace Impurities - Mercury (Hg) ≤0.5 ppb
Trace Impurities - Molybdenum (Mo) ≤10.0 ppb
Trace Impurities - Nickel (Ni) ≤2.0 ppb
Trace Impurities - Niobium (Nb) ≤10.0 ppb
Trace Impurities - Potassium (K) ≤500.0 ppb
Trace Impurities - Selenium (Se) ≤50.0 ppb
Trace Impurities - Silicon (Si) ≤100.0 ppb
Trace Impurities - Silver (Ag) ≤1.0 ppb
Trace Impurities - Sodium (Na) ≤500.0 ppb
Trace Impurities - Strontium (Sr) ≤5.0 ppb
Trace Impurities - Tantalum (Ta) ≤10.0 ppb
Trace Impurities - Thallium (Tl) ≤20.0 ppb
Trace Impurities - Tin (Sn) ≤5.0 ppb
Trace Impurities - Titanium (Ti) ≤10.0 ppb
Trace Impurities - Vanadium (V) ≤10.0 ppb
Trace Impurities - Zinc (Zn) ≤5.0 ppb
Trace Impurities - Zirconium (Zr) ≤10.0 ppb

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR