Sodium hypochlorite 5% Chlorine_available aqueous solution, BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
M.O.C. Category N
9416-01
9416-03
CAJT9416-1CS
33452.85
CAD
CAJT9416-1
CAJT9416-3
JT9416-1
JT9416-3
Sodium hypochlorite 5% Chlorine_available aqueous solution, BAKER ANALYZED®, J.T.Baker®
Sodium hypochlorite
Formula:
ClNaO MW: 74.44 g/mol Melting Pt: -6 Storage Temperature: Refrigerator |
MDL Number:
MFCD00011120 CAS Number: 7681-52-9 EINECS: 231-668-3 Merck Index: 13,08702 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Appearance (Clear, light yellow liquid) | Passes Test |
Available Chlorine (when packaged) | 5.0 - 6.0 % |
Phosphate (PO₄) | ≤0.0005 % |
Calcium (Ca) | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...