Sodium hydroxide 0.2 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Caustic soda

Management of Change (MOC) category = NI

BAKR5665-07 BAKR5665-02 BAKR5665-03
CAJT5665-7EA 583.84 CAD
CAJT5665-7 CAJT5665-2 CAJT5665-3
Sodium hydroxide 0.2 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium hydroxide

Standardization at 25 °C traceable to NIST Standard Reference Material.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Protect from air to avoid absorption of carbon dioxide.
Normality 0.1995 - 0.2005
Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR