Sulfuric acid 1 N aqueous solution, BAKER ANALYZED® Volumetric, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


M.O.C. Category NI

BAKR5642-02 BAKR5642-03
CAJT5642-2CS 598.16 CAD
CAJT5642-2 CAJT5642-3
Sulfuric acid 1 N aqueous solution, BAKER ANALYZED® Volumetric, J.T.Baker®
Sulfuric acid

The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Normality 0.995 - 1.005
Chloride (Cl) ≤1 ppm
Phosphate (PO₄) ≤1 ppm
Heavy Metals (as Pb) ≤0.3 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR