≥98% (dried basis) USP, Multi-Compendial endotoxin-free

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

Management of Change (MOC) category = HR

BAKR3250-06 BAKR3250-07 BAKR3250-05 3250-60 3250-65 BAKR3250-60 BAKR3250-65
CAJT3250-6EA 2623.59 CAD
CAJT3250-6 CAJT3250-7 CAJT3250-1 CAJT3250-5 JT3250-60 JT3250-65 CAJT3250-60 CAJT3250-65
≥98% (dried basis) USP, Multi-Compendial endotoxin-free
di-Potassium hydrogen phosphate

GMP manufactured.


  • GMP manufactured product

Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (K₂HPO₄) (dried basis) 98.0 - 100.5 %
USP - Arsenic (As) ≤ 3 ppm
USP - Carbonate (CO₃) Passes Test
USP - Chloride (Cl) ≤ 0.03 %
USP - Endotoxin Concentration (2.5 EU/g max.) Passes Test
USP - Fluoride (F) ≤ 0.001 %
USP - Identification A (USP) Passes Test
USP - Identification B (USP) Passes Test
USP - Insoluble Substances ≤ 0.2 %
USP - Iron (Fe) ≤ 0.003 %
USP - Loss on Drying ≤ 1.0 %
USP - Mono- or Tri- Basic Salt Passes Test
USP - pH (1 in 20) 8.5 - 9.6
USP - Sodium (Na) Passes Test
USP - Sulfate (SO₄) ≤ 0.1 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Assay (K₂HPO₄) (dried basis) 98.0 - 101.0 %
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying ≤ 2.0 %
EP/BP - Monopotassium phosphate ≤ 0.025
EP/BP - Reducing Substances Passes Test
EP/BP - Sodium (Na) ≤ 0.1 %
EP/BP - Sulfate (SO₄) ≤ 0.1 %
Color (APHA), For Information Only
Aluminum (Al), For Information Only
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Preserve in well closed container. Store at ambient temperature.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR