Sodium thiosulfate 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = NI

BAKR5637-07 BAKR5637-03 5637-02
CAJT5637-7EA 475.07 CAD
CAJT5637-7 CAJT5637-2 CAJT5637-3 JT5637-2
Sodium thiosulfate 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium thiosulphate

J.T.Baker® volumetric and analytical solutions. These solutions are suitable for use in ACS, USP and NF compendial methods and general laboratory applications. 

Formula: Na₂S₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005
Sulfate and Sulfite (as SO₄) ≤0.01 %
ACS - Heavy Metals (as Pb) ≤1 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR