Sodium hydroxide 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = NI
BAKR5636-07
BAKR5636-02
BAKR5636-03
CAJT5636-7EA
527.28
CAD
CAJT5636-7
CAJT5636-2
CAJT5636-3
Sodium hydroxide 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium hydroxide
Standardization at 25 °C traceable to NIST Standard Reference Material.
Formula:
NaOH Density: 1 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Protect from air to avoid absorption of carbon dioxide. | |
Normality | 0.0995 - 0.1005 |
Chloride (Cl) | ≤5 ppm |
ACS - Heavy Metals (as Pb) | ≤1 ppm |
Trace Impurities - Iron (Fe) | ≤0.5 ppm |
Standardization at 25°C traceable to NIST Standard Reference Material. | |
Filtered through a 0.2 micron filter. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...