Sodium hydroxide 1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Caustic soda

Management of Change (MOC) category = NI

BAKR5635-02 BAKR5635-03 5635-07
CAJT5635-2CS 581.88 CAD
CAJT5635-2 CAJT5635-3 JT5635-7
Sodium hydroxide 1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium hydroxide

Standardization at 25 °C traceable to NIST Standard Reference Material.

Formula: NaOH
Density: 1.04 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Protect from air to avoid absorption of carbon dioxide.
Appearance (Clear, colorless liquid) Passes Test
Normality (eq/L) 0.995 - 1.005
Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR