Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

Management of Change (MOC) category = NI

BAKR5630-02 BAKR5630-03
CAJT5630-2CS 4039.95 CAD
CAJT5630-2 CAJT5630-3
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Silver nitrate

J.T.Baker® volumetric and analytical solutions. These solutions are suitable for use in ACS, USP and NF compendial methods and general laboratory applications. 

Formula: AgNO₃
MW: 169.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.095 - 0.105
pH at 25°C 3.0 - 6.0
Chloride (Cl) ≤1 ppm
Sulfate (SO₄) ≤1 ppm
Appearance Passes Test
Standardization at 25°C traceable to NIST Standard Reference Material.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR