Sodium hydroxide 10 N aqueous solution, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Caustic soda

M.O.C. Category R

BAKR5000-07 BAKR5000-09 BAKR5000-02 BAKR5000-03 BAKR5000-27 BAKR5000-30
CAJT5000-7EA 4195.55 CAD
CAJT5000-7 CAJT5000-9 CAJT5000-02 CAJT5000-3 CAJT5000-27 CAJT5000-30
Sodium hydroxide 10 N aqueous solution, Biotech Reagent, J.T.Baker®
Sodium hydroxide

Clear, colorless liquid. Made from USP purified water and NF (multicompendial) sodium hydroxide pellets that meet Ph.Eur., BP, and JP chemical specifications. Filtered through a 0.2µm filter. Completely miscible with water. GMP manufactured product.


Made from USP purified water and NF (multicompendial) sodium hydroxide pellets that meet Ph. Eur., BP, and JP chemical specifications.
Filtered through a 0.2 µm filter. Completely miscible with water. GMP manufactured product.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

GMP Manufactured Product
IMPORTANT: Material will freeze if stored below 41 °F (5°C).
Protect from air to avoid absorption of carbon dioxide.
CAUTION: For Manufacturing, processing or repackaging
Appearance (Clear, colorless liquid) Passes Test
Normality 9.9 - 10.1
Sodium Carbonate (Na₂CO₃) ≤0.6 %
Chloride (Cl) ≤20 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤2 ppm
Endotoxin Concentration (EU/mL) ≤2.0
Made from U.S.P. Purified Water and Sodium Hydroxide pellets, N.F. (Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
IMPORTANT: Material will freeze if stored below 41°F (5°C).
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR