Potassium dihydrogen phosphate 98 Dried, crystals NF, multi-compendial

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

M.O.C. Category R

BAKR3248-07 BAKR3248-01 BAKR3248-05 BAKR3248-15 BAKR3248-60 BAKR3248-F7
CAJT3248-7EA 3304.54 CAD
CAJT3248-7 CAJT3248-1 CAJT3248-5 CAJT3248-15 CAJT3248-60 CAJT3248-F7
Potassium dihydrogen phosphate 98 Dried, crystals NF, multi-compendial
Potassium dihydrogen phosphate

Maximize the stability of your pharmaceutical formulations and get predictable pH control with reliable, proven buffering agents from Avantor™ Performance Materials.


  • GMP manufactured product


No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Certifications: Meets NF requirements and EP and BP chemical specifications.


Caution: Store at 20° to 25°C (68° to 77°F), excursion permitted to 15° to 30°C (59° to 86°F) [see USP controlled Room Temperature].

Formula: KH₂PO₄
MW: 136.09 g/mol
Boiling Pt: 100
Melting Pt: 253
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Order Now

Specification Test Results

Meets N.F. Requirements
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
GMP Manufactured Product
Preserve in Tight Containers
Preserve in well-closed containers.
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (KH₂PO₄) (dried basis) 98.0 - 100.5 %
NF - Arsenic (As) ≤ 3 ppm
NF - Endotoxin Concentration (2.5 EU/g max.) Passes Test
NF - Fluoride (F) ≤ 0.001 %
NF - Identification Passes Test
NF - Insoluble Substances ≤ 0.2 %
NF - Lead (Pb) ≤ 2 ppm
NF - Loss on Drying at 105°C ≤ 1.0 %
EP/BP - Assay (KH₂PO₄) (dried basis) 98.0 - 100.5 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying at 125 - 130°C ≤ 2.0 %
EP/BP - Reducing Substances Passes Test
EP/BP - pH 4.2 - 4.5
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Arsenic (As) ≤ 2 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR