Potassium peroxodisulfate 99,0, BAKER INSTRA-ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: di-Potassium peroxodisulphate, Dipotassium peroxodisulfate, Peroxydisulfuric acid dipotassium salt, Potassium persulfate, di-Potassium peroxodisulfate, Peroxydisulphuric acid dipotassium salt, Dipotassium peroxodisulphate, Potassium persulphate

M.O.C. Category N

3239-01 3239-05
JT3239-1CS 1388.36 CAD
JT3239-1 JT3239-5
Potassium peroxodisulfate 99,0, BAKER INSTRA-ANALYZED® ACS
Potassium peroxodisulphate

For Kjeldahl nitrogen, trace phosphate, and mercury determinations. Lot analysis on label.

Formula: K₂S₂O₈
MW: 270.32 g/mol
Melting Pt: 100
Density: 2.477 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011386
CAS Number: 7727-21-1
EINECS: 231-781-8
UN: 1492
ADR: 5.1 III
Merck Index: 13,07741

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (K₂S₂O₈) ≥99.0 %
Insoluble Matter ≤0.005 %
Chlorine Compounds (as Cl) ≤0.001 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤5 ppm
Trace Impurities - Manganese (Mn) ≤2.000 ppm
Trace Impurities - Mercury (Hg), For Information Only
Phosphate (PO₄) ≤2.5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR