Potassium peroxodisulfate ≥99% (by iodometry), BAKER ANALYZED®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: di-Potassium peroxodisulphate, Dipotassium peroxodisulfate, Peroxydisulfuric acid dipotassium salt, Potassium persulfate, di-Potassium peroxodisulfate, Peroxydisulphuric acid dipotassium salt, Dipotassium peroxodisulphate, Potassium persulphate

Management of Change (MOC) category = N

BAKR3238-08 BAKR3238-01 BAKR3238-05
CAJT3238-8EA 6556.67 CAD
CAJT3238-8 CAJT3238-1 CAJT3238-5
Potassium peroxodisulfate ≥99% (by iodometry), BAKER ANALYZED®
Potassium peroxodisulphate

White powder. Lot analysis on label.

Formula: K₂S₂O₈
Melting Pt: 100 °C
Density: 2.477 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011386
CAS Number: 7727-21-1
EINECS: 231-781-8
UN: 1492
ADR: 5.1,III
Merck Index: 13,07741

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (K₂S₂O₈) (by gravimetry) ≥99.0 %
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Manganese (Mn) ≤2.000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR