D-(+)-Sucrose ≥99.9% (by HPLC), Ultrapure Bioreagent for density gradient ultracentrifugation, for molecular biology, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
|
Synonyms:
β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
Management of Change (MOC) category = N
BAKR4097-04
BAKR4097-06
BAKR4097-07
BAKR4097-01
CAJT4097-4EA
248.44
CAD
CAJT4097-4
CAJT4097-6
CAJT4097-7
CAJT4097-01
D-(+)-Sucrose ≥99.9% (by HPLC), Ultrapure Bioreagent for density gradient ultracentrifugation, for molecular biology, J.T.Baker®
D(+)-Sucrose
Sucrose for Density Gradient Centrifugation and Molecular Biology Applications.
Formula:
C₁₂H₂₂O₁₁ Melting Pt: 169…170 °C Density: 1.29 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006626 CAS Number: 57-50-1 EINECS: 200-334-9 Merck Index: 13,08966 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
DNase Activity (None Detected) | Passes Test |
RNase Activity | Passes Test |
Protease Activity | Passes Test |
Clarity and Color of Solution | Passes Test |
Clarity of Solution | Passes Test |
Glucose | ≤0.1 % |
Total of Copper, Iron and Lead | ≤5 ppm |
Assay (C₁₂H₂₂O₁₁) (by HPLC) | ≥99.9 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...