Potassium periodate 99.8-100.3% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Potassium tetroxoiodate (VII), Potassium metaperiodate

Management of Change (MOC) category = N

BAKR3224-04 BAKR3224-01
CAJT3224-4EA 517.97 CAD
CAJT3224-4 CAJT3224-1
Potassium periodate 99.8-100.3% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®
Potassium periodate

Lot analysis on label.

Formula: KlO₄
MW: 230 g/mol
Melting Pt: 576…588 °C
Density: 3.62 g/cm³ (25 °C)
MDL Number: MFCD00011407
CAS Number: 7790-21-8
EINECS: 232-196-0
UN: 1479
ADR: 5.1,II
Merck Index: 13,07739

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (KIO₄) 99.8 - 100.3 %
Insoluble Matter ≤0.01 %
Other Halogens (as Cl) ≤0.01 %
Trace Impurities - Manganese (Mn) ≤0.500 ppm
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR