Potassium hydroxide 45% ∼11.7 N, BAKER ANALYZED®

Supplier: VWR International
Danger

Synonyms: Potash caustic, Caustic potash

Management of Change (MOC) category = N

CAJT3143-7 BAKR3143-01 BAKR3143-03 3143-R
CAJT3143-7EA 2071.06 CAD
CAJT3143-7 CAJT3143-R CAJT3143-1 CAJT3143-3 JT3143-R
Potassium hydroxide 45% ∼11.7 N, BAKER ANALYZED®
Potassium hydroxide

Lot analysis on label.

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (KOH) 45.0 - 46.5 %
Potassium Carbonate (K₂CO₃) ≤0.2 %
Chloride (Cl) ≤0.003 %
Sulfate (SO₄) ≤0.001 %
Ammonium Hydroxide Precipitate ≤0.005 %
Sodium (Na) ≤0.02 %
Nitrogen Compounds (as N) ≤5 ppm
Phosphate (PO₄) ≤2 ppm
Heavy Metals (as Ag) ≤5 ppm
Trace Impurities - Copper (Cu) ≤0.050 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Trace Impurities - Nickel (Ni) ≤5.000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR