Sodium dodecyl sulfate (SDS) ≥99%, Ultrapure Bioreagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
Management of Change (MOC) category = N
BAKR4095-02
BAKR4095-07
BAKR4095-04
CAJT4095-2EA
1054.52
CAD
CAJT4095-2
CAJT4095-5
CAJT4095-7
CAJT4095-4
Sodium dodecyl sulfate (SDS) ≥99%, Ultrapure Bioreagent, J.T.Baker®
Sodium dodecyl sulphate (SDS)
For electrophoresis and protein solubilization.
- Lot analysis on label
Formula:
CH₃(CH₂)₁₁OSO₃Na MW: 288.38 g/mol Melting Pt: 204…207 °C Density: 0.40 g/cm³ Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 2926 ADR: 4.1 (6.1) II Merck Index: 14,08636 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Appearance | Passes Test |
Assay (as C₁₂H₂₅OSO₃Na) | ≥99 % |
Absorbance of a 3% w/v Aqueous Solution at 280 nm (1-cm path) | ≤0.1 |
pH of 1% Solution at 25°C | 5.0 - 8.0 |
Chloride (Cl) | ≤0.03 % |
Nitrogen (N) | ≤5 ppm |
Phosphate (PO₄) | ≤1 ppm |
Heavy Metals (as Pb) | ≤2 ppm |
Trace Impurities - Iron (Fe) | ≤1 ppm |
DNase Activity (None Detected) | Passes Test |
RNase Activity | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...