Nalgene® Volumetric Flasks with Screw Cap, Polymethylpentene, Thermo Scientific
Supplier: Thermo Fisher Scientific
4001-0050 4001-0100 4001-0250 4001-0500 4001-1000
29615-255CS 908.41 CAD
29615-255 29615-277 29615-299 29615-313 29615-335
Nalgene® Volumetric Flasks with Screw Cap, Polymethylpentene, Thermo Scientific
Flasks Volumetric Flasks
Transparent polymethylpentene flasks are lighter in weight than glass and break resistant to protect against leakage and contamination
Each flask is individually hand calibrated to contain and to deliver at 20°C (68°F). Flasks contain a permanent color graduation line and oval marking spot for ease in reading fluid levels. Autoclavable, leakproof, and graduated.
Certifications: Flasks meet ASTM E288 and ISO 384 Class B tolerances for consistent quality and dependability.